1,2,3-Tribromopropane 98%

Warning

L03752-22 L03752-14 L03752-14 L03752-22
AAAL03752-22EA 45.05 USD
AAAL03752-22 CAAAAL03752-22 AAAL03752-14 CAAAAL03752-14
1,2,3-Tribromopropane 98%
1,2,3-Tribromopropane
Formula: BrH₂CBrCHCH₂Br
MW: 280.78 g/mol
Boiling Pt: 220 °C
Melting Pt: 16…17 °C
Density: 2.398
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017884
CAS Number: 96-11-7
EINECS: 202-478-8
UN: 2810
ADR: "6.1",III
Merck Index: 12,09745

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR