Iodoacetonitrile 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Iodoacetonitrile

L03634-18 L03634-18 L03634-09 L03634-09
AAAL03634-09EA 59.04 USD
AAAL03634-09 CAAAAL03634-09 AAAL03634-18 CAAAAL03634-18
Iodoacetonitrile 97%
Iodoacetonitrile
Formula: C₂H₂IN
MW: 166.95 g/mol
Boiling Pt: 70…72 °C/10 mm
Density: 2.306
Flash Pt: 86 °C(186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001886
CAS Number: 624-75-9
EINECS: 210-860-0
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR