Ethyl trifluoromethanesulfonate 99%

Danger

L03383-06 L03383-06
AAAL03383-06EA 67.35 USD
CAAAAL03383-06 AAAL03383-06
Ethyl trifluoromethanesulfonate 99%
Ethyl trifluoromethanesulphonate
Formula: C₃H₅F₃O₃S
MW: 178.13 g/mol
Boiling Pt: 115…118 °C
Density: 1.374
Flash Pt: 35 °C(95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000410
CAS Number: 425-75-2
EINECS: 207-037-3
UN: 2920
ADR: "8",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR