2,5-Dibromo-α,α,α-trifluorotoluene 98%

Warning

Synonyms: 1,4-Dibromo-2-(trifluoromethyl)benzene2,5-Dibromobenzotrifluoride

L03378-06 L03378-14 L03378-14 L03378-06
AAAL03378-14EA 251.97 USD
CAAAAL03378-06 CAAAAL03378-14 AAAL03378-14 AAAL03378-06
2,5-Dibromo-α,α,α-trifluorotoluene 98%
2,5-Dibromo-α,α,α-trifluorotoluene
Formula: C₇H₃Br₂F₃
MW: 303.9 g/mol
Boiling Pt: 210 °C
Melting Pt: 47…51 °C
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013554
CAS Number: 7657-09-2
EINECS: 231-620-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR