2-Amino-9,10-anthraquinone tech. 90%, Technical Grade

Warning

Synonyms: 2-Aminoanthraquinone

L03368-36 L03368-22
CAAAAL03368-36 CAAAAL03368-22
2-Amino-9,10-anthraquinone tech. 90%, Technical Grade
2-Amino-9,10-anthraquinone
Formula: C₁₄H₉NO₂
MW: 223.23 g/mol
Melting Pt: ca 293 °C (dec.)
Flash Pt: 283 °C(541 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001233
CAS Number: 117-79-3
EINECS: 204-208-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR