2-Hydroxy-p-toluic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylsalicylic acidm-Cresotic acidm-Homosalicyclic acid2-Hydroxy-4-methylbenzoic acid

L03367-14 L03367-22 L03367-22 L03367-14
AAAL03367-22EA 103.99 USD
AAAL03367-22 CAAAAL03367-22 AAAL03367-14 CAAAAL03367-14
2-Hydroxy-p-toluic acid 99%
2-Hydroxy-p-toluic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00002454
CAS Number: 50-85-1
EINECS: 200-068-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR