2-Pentyne 98+%

Supplier: Thermo Scientific Chemicals
Danger

L02973-14 L02973-14
AAAL02973-14EA 138.99 USD
CAAAAL02973-14 AAAL02973-14
2-Pentyne 98+%
2-Pentyne
Formula: C₅H₈
MW: 68.12 g/mol
Boiling Pt: 55…56 °C
Melting Pt: –109 °C
Density: 0.710
Flash Pt: -20 °C(-4 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009380
CAS Number: 627-21-4
EINECS: 210-989-2
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR