1-Pentadecyne 98%

Warning

L02540-04 L02540-09 L02540-04 L02540-09
AAAL02540-09EA 215.78 USD
AAAL02540-09 CAAAAL02540-09 AAAL02540-04 CAAAAL02540-04
1-Pentadecyne 98%
1-Pentadecyne
Formula: C₁₅H₂₈
MW: 208.39 g/mol
Boiling Pt: 129…131 °C/10 mm
Melting Pt: 10 °C
Density: 0.793
Flash Pt: >90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015081
CAS Number: 765-13-9
EINECS: 212-140-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR