4-Iodo-o-toluidine 98+%

Warning

Synonyms: 2-Amino-5-iodotoluene4-Iodo-2-methylaniline

L02306-06 L02306-14 L02306-06 L02306-14
AAAL02306-06EA 74.61 USD
AAAL02306-06 AAAL02306-14 CAAAAL02306-14 CAAAAL02306-06
4-Iodo-o-toluidine 98+%
4-Iodo-o-toluidine
Formula: C₇H₈IN
MW: 233.05 g/mol
Melting Pt: 84…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00025299
CAS Number: 13194-68-8
EINECS: 236-154-2
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR