Dibenzosuberenol 96%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Dibenzo[b,f]cycloheptenolDibenzo[b,f]cyclohepten-1-ol1-Hydroxydibenzo[b,f]cycloheptene5-Dibenzosuberenol5H-Dibenzo[a,d][7]annulen-5-ol

L02062-14 L02062-06 L02062-14 L02062-06
AAAL02062-06EA 60.29 USD
CAAAAL02062-14 AAAL02062-06 AAAL02062-14 CAAAAL02062-06
Dibenzosuberenol 96%
Dibenzosuberenol
Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00003586
CAS Number: 10354-00-4
EINECS: 233-774-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR