2-Fluoroethanol 95%

Danger

L01735-06 L01735-14 L01735-06 L01735-14
AAAL01735-06EA 92.64 USD
AAAL01735-06 CAAAAL01735-06 AAAL01735-14 CAAAAL01735-14
2-Fluoroethanol 95%
2-Fluoroethanol
Formula: C₂H₅FO
MW: 64.06 g/mol
Boiling Pt: 102…104 °C
Melting Pt: –27 °C
Density: 1.104
Flash Pt: 31 °C(87 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002828
CAS Number: 371-62-0
EINECS: 206-740-2
UN: 3383
ADR: "6.1",I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR