α-Bromo-2,6-dichlorotoluene 97%

Danger

Synonyms: alpha-Bromo-2,6-dichlorotoluene2,6-Dichlorobenzyl bromide

L01710-14 L01710-22 L01710-22 L01710-14
AAAL01710-22EA 161.29 USD
CAAAAL01710-22 AAAL01710-22 CAAAAL01710-14 AAAL01710-14
α-Bromo-2,6-dichlorotoluene 97%
α-Bromo-2,6-dichlorotoluene
Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
Melting Pt: 54…57 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000577
CAS Number: 20443-98-5
EINECS: 243-827-4
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR