Iodocyclopentane 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cyclopentyl iodide

Stabilization: Stabilised with copper

L01522-06 L01522-06 L01522-14 L01522-14
AAAL01522-14EA 151.99 USD
AAAL01522-14 CAAAAL01522-14 CAAAAL01522-06 AAAL01522-06
Iodocyclopentane 97% stabilized
Iodocyclopentane
Formula: C₅H₉I
MW: 196.03 g/mol
Boiling Pt: 166…167 °C
Density: 1.695
Flash Pt: 59 °C(138 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039446
CAS Number: 1556-18-9
EINECS: 216-311-1
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR