5-Methylanthranil acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Amino-m-toluic acid2-Amino-5-methylbenzoic acid

L01513-14 L01513-03 L01513-14 L01513-06 L01513-03 L01513-06
AAAL01513-06EA 136.99 USD
AAAL01513-06 CAAAAL01513-03 CAAAAL01513-14 AAAL01513-14 AAAL01513-03 CAAAAL01513-06
5-Methylanthranil acid 97%
5-Methylanthranil acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 317 °C (1013 hPa)
Melting Pt: ca 175 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007909
CAS Number: 2941-78-8
EINECS: 220-932-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR