3-Nitrobiphenyl 98+%

Warning

Synonyms: 3-Nitro-1,1'-biphenyl

L01246-04 L01246-09 L01246-09 L01246-04
AAAL01246-09EA 139.57 USD
CAAAAL01246-04 CAAAAL01246-09 AAAL01246-09 AAAL01246-04
3-Nitrobiphenyl 98+%
3-Nitrobiphenyl
Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
Boiling Pt: 227 °C/35 mm
Melting Pt: 58…60 °C
Density: 1.305 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007245
CAS Number: 2113-58-8
EINECS: 218-305-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR