(4-Iodophenoxy)acetic acid 98+%

Warning

L01196-06 L01196-06
AAAL01196-06EA 58.05 USD
AAAL01196-06 CAAAAL01196-06
(4-Iodophenoxy)acetic acid 98+%
(4-Iodophenoxy)acetic acid
Formula: (C₆H₄I)OCH₂COOH
MW: 278.05 g/mol
Boiling Pt: 342 °C (1013 hPa)
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00014359
CAS Number: 1878-94-0
EINECS: 217-531-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR