(2,5-Xylyloxy)acetic acid 98+%

Warning

Synonyms: 2-(2,5-Dimethylphenoxy)acetic acid(2,5-Dimethylphenoxy)acetic acid

L01176-14 L01176-06 L01176-14 L01176-06
AAAL01176-06EA 51.34 USD
CAAAAL01176-14 AAAL01176-06 AAAL01176-14 CAAAAL01176-06
(2,5-Xylyloxy)acetic acid 98+%
(2,5-Xylyloxy)acetic acid
Formula: (C₆H₃(CH₃)₂)OCH₂COOH
MW: 180.2 g/mol
Melting Pt: 114…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00014356
CAS Number: 7356-41-4
EINECS: 230-887-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR