6-Methyl-5-nitroquinoline 97%

Warning

L00807-06 L00807-03 L00807-06 L00807-03
AAAL00807-03EA 33.61 USD
AAAL00807-03 CAAAAL00807-06 AAAL00807-06 CAAAAL00807-03
6-Methyl-5-nitroquinoline 97%
6-Methyl-5-nitroquinoline
Formula: C₁₀H₈N₂O₂
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00024021
CAS Number: 23141-61-9
EINECS: 245-447-4
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR