Benzyltripropylammonium chloride 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Benzyl-N,N-dipropylpropan-1-aminium chloride
L00675-09
L00675-09
AAAL00675-09EA
55.89
USD
AAAL00675-09
CAAAAL00675-09
Benzyltripropylammonium chloride 96%
Benzyltripropylammonium chloride
Formula:
C₁₆H₂₈ClN MW: 269,85 g/mol Melting Pt: ca 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051869 CAS Number: 5197-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...