N,N-Dimethyl-3-nitroaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

L00401-06 L00401-14 L00401-06 L00401-14
AAAL00401-06EA 41.79 USD
AAAL00401-06 CAAAAL00401-06 CAAAAL00401-14 AAAL00401-14
N,N-Dimethyl-3-nitroaniline 98%
N,N-Dimethyl-3-nitroaniline
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00007236
CAS Number: 619-31-8
EINECS: 210-590-3
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR