1-Vinylnaphthalene 95% stabilized

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

H51043-03 H51043-06
AAH51043-03EA 79.59 USD
AAH51043-03 AAH51043-06
1-Vinylnaphthalene 95% stabilized
1-Vinylnaphthalene
Formula: C₁₂H₁₀
MW: 154.21 g/mol
Boiling Pt: 135…138 °C
Melting Pt: 21 °C
Density: 1.04
Storage Temperature: Refrigerator
MDL Number: MFCD00075766
CAS Number: 826-74-4
EINECS: 212-560-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR