2,4-Dimethylphenylacetic acid 98%

Warning

Synonyms: 2,4-Xylylacetic acid(2,4-Dimethylphenyl)acetic acid

H50284-03 H50284-03 H50284-MD
AAAH50284-03EA 247.46 USD
CAAAAH50284-03 CAAAAH50284-MD AAAH50284-03
2,4-Dimethylphenylacetic acid 98%
2,4-Dimethylphenylacetic acid
Formula: (C₆H₃(CH₃)₂)CH₂COOH
MW: 164.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02664685
CAS Number: 6331-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR