2-Fluoro-3-iodopyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H50146-MD H50146-MD H50146-03 H50146-03
AAAH50146-03EA 164.99 USD
AAAH50146-03 CAAAAH50146-03 AAAH50146-MD CAAAAH50146-MD
2-Fluoro-3-iodopyridine 97%
2-Fluoro-3-iodopyridine
Formula: C₅H₃FIN
MW: 222.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03095287
CAS Number: 113975-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR