2-Ethoxy-5-pyridineboronic acid pinacol ester 99%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 2-Ethoxypyridine-5-boronic acid pinacol ester2-Ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

H50137-03 H50137-03 H50137-MD H50137-MD
AAAH50137-MDEA 62.87 USD
AAAH50137-MD CAAAAH50137-03 AAAH50137-03 CAAAAH50137-MD
2-Ethoxy-5-pyridineboronic acid pinacol ester 99%
2-Ethoxy-5-pyridineboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₃
MW: 249.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781182
CAS Number: 1072945-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR