2-Bromo-5-pyridineboronic acid pinacol ester 98%

Warning

Synonyms: 2-Bromopyridine-5-boronic acid pinacol ester2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

H50072-03 H50072-03 H50072-06 H50072-06
AAAH50072-03EA 197.01 USD
AAAH50072-03 CAAAAH50072-03 CAAAAH50072-06 AAAH50072-06
2-Bromo-5-pyridineboronic acid pinacol ester 98%
2-Bromo-5-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BBrNO₂
MW: 283.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412794
CAS Number: 214360-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR