2-Chloro-5-pyridineboronic acid pinacol ester 98%

Warning

Synonyms: 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine2-Chloropyridine-5-boronic acid Pinacol ester2-(6-Chloro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H50071-06 H50071-03 H50071-06 H50071-03
AAAH50071-03EA 95.35 USD
CAAAAH50071-03 CAAAAH50071-06 AAAH50071-03 AAAH50071-06
2-Chloro-5-pyridineboronic acid pinacol ester 98%
2-Chloro-5-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BClNO₂
MW: 239.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03412793
CAS Number: 444120-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR