2-Chloro-4-pyridineboronic acid pinacol ester 98%

Warning

Synonyms: 2-(2-Chloro-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane2-Chloropyridine-4-boronic acid Pinacol ester2-Chloropyridine-4-boronic acid pinacol ester2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

H50070-03 H50070-06 H50070-06 H50070-03
AAAH50070-03EA 229.98 USD
CAAAAH50070-03 AAAH50070-03 CAAAAH50070-06 AAAH50070-06
2-Chloro-4-pyridineboronic acid pinacol ester 98%
2-Chloro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BClNO₂
MW: 239.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04039870
CAS Number: 458532-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR