4-Aminonicotinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-3-pyridinecarboxylic acid

H50032-03 H50032-03 H50032-06 H50032-06
AAAH50032-03EA 30.19 USD
CAAAAH50032-03 AAAH50032-03 CAAAAH50032-06 AAAH50032-06
4-Aminonicotinic acid 98%
4-Aminonicotinic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00234183
CAS Number: 7418-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR