N,N-Dibenzylaniline 99%

Supplier: Thermo Scientific Chemicals
Warning

H32091-22 H32091-14
AAH32091-22EA 142.99 USD
AAH32091-22 AAH32091-14
N,N-Dibenzylaniline 99%
N,N-Dibenzylaniline
Formula: (C₆H₅)N(CH₂C₆H₅)₂
MW: 273.38 g/mol
Boiling Pt: 180 °C/1 mm
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00022015
CAS Number: 91-73-6
EINECS: 202-093-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR