(3-Methoxy-5-(trifluoromethyl)phenyl)acetic acid 97%

Supplier: Alfa Aesar
Warning

H31985-MD H31985-03
AAH31985-MDEA 101.65 USD
AAH31985-MD AAH31985-03
(3-Methoxy-5-(trifluoromethyl)phenyl)acetic acid 97%
(3-Methoxy-5-(trifluoromethyl)phenyl)acetic acid
Formula: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD09025416
CAS Number: 916421-04-0
EINECS: 000-000-0
UN: 0000

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR