5-Bromoindol-3-ylacetic acid 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-Bromoindole-3-acetic acid2-(5-Bromo-1H-indol-3-yl)acetic acid

H29217-03 H29217-MD
AAH29217-03EA 211 USD
AAH29217-03 AAH29217-MD
5-Bromoindol-3-ylacetic acid 97%
5-Bromoindol-3-ylacetic acid
Formula: C₁₀H₈BrNO₂
MW: 254.08 g/mol
Melting Pt: 144…145 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005637
CAS Number: 40432-84-6
EINECS: 254-917-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR