1-Isoquinolinol 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Isocarbostyril
, 1-Hydroxyisoquinoline
H29124-09
H29124-03
AAH29124-03EA
88.89
USD
AAH29124-03
AAH29124-09
1-Isoquinolinol 97%
1-Isoquinolinol
Formula:
C₉H₇NO MW: 145.16 g/mol Melting Pt: 212…215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006899 CAS Number: 491-30-5 EINECS: 207-732-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...