Potassium-3-iodophenyltrifluoroborate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium 3-iodophenyltrifluoroborate

H29009-06 H29009-03
AAH29009-06EA 205.99 USD
AAH29009-06 AAH29009-03
Potassium-3-iodophenyltrifluoroborate 96%
Potassium 3-iodophenyltrifluoroborate
Formula: C₆H₄BIF₃K
MW: 309.91 g/mol
Melting Pt: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD09800739
CAS Number: 1189097-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR