4-Cyanophenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
, 4-Cyanobenzeneboronic acid pinacol ester
H28208-03
H28208-06
AAH28208-06EA
238.99
USD
AAH28208-06
AAH28208-03
4-Cyanophenylboronic acid pinacol ester 97%
4-Cyanophenylboronic acid pinacol ester
Formula:
C₁₃H₁₆BNO₂ MW: 229.09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03093897 CAS Number: 171364-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...