2,7-Dimethoxynaphthalene 98%

H27971-14 H27971-06
AAH27971-14EA 181.79 USD
AAH27971-14 AAH27971-06
2,7-Dimethoxynaphthalene 98%
2,7-Dimethoxynaphthalene
Formula: C₁₂H₁₂O₂
MW: 188.23 g/mol
Melting Pt: 137…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00004064
CAS Number: 3469-26-9
EINECS: 222-433-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR