6-Bromonicotinic acid 97%

Warning

Synonyms: 6-Bromopyridine-3-carboxylic acid2-Bromo-5-pyridinecarboxylic acid6-Bromo-3-pyridinecarboxylic acid

H27926-14 H27926-06
AAH27926-14EA 421.99 USD
AAH27926-14 AAH27926-06
6-Bromonicotinic acid 97%
6-Bromonicotinic acid
Formula: C₆H₄BrNO₂
Melting Pt: 179…188 °C
Storage Temperature: Ambient
MDL Number: MFCD01927100
CAS Number: 6311-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR