4-Hydroxy-3-methylbenzoic acid hydrate 97%

Warning

Synonyms: 4-Hydroxy-m-toluic acid

H27792-09 H27792-03
AAH27792-03EA 26.64 USD
AAH27792-03 AAH27792-09
4-Hydroxy-3-methylbenzoic acid hydrate 97%
4-Hydroxy-3-methylbenzoic acid hydrate
Formula: C₈H₈O₃·xH₂O
MW: 152.15 g/mol (anhy)
Melting Pt: 173…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00270105
CAS Number: 499-76-3
EINECS: 207-890-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR