2,5-Dichloro-4-pyridineboronic acid pinacol ester 95%

Warning

H27699-03 H27699-MD
AAH27699-MDEA 156.51 USD
AAH27699-MD AAH27699-03
2,5-Dichloro-4-pyridineboronic acid pinacol ester 95%
2,5-Dichloro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₄BCl₂NO₂
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD06798256
CAS Number: 1073353-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR