2,6-Dichloro-3-pyridineboronic acid pinacol ester 95%

Warning

H27605-MD H27605-03
AAH27605-03EA 231.99 USD
AAH27605-03 AAH27605-MD
2,6-Dichloro-3-pyridineboronic acid pinacol ester 95%
2,6-Dichloro-3-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₄BCl₂NO₂
Melting Pt: 51…53 °C
Storage Temperature: Ambient
MDL Number: MFCD06798261
CAS Number: 1073371-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR