2-Bromo-4-methylbenzaldehyde 95%

Supplier: Thermo Scientific Chemicals
Warning

H27558-03
AAH27558-03EA 70.99 USD
AAH27558-03
2-Bromo-4-methylbenzaldehyde 95%
2-Bromo-4-methylbenzaldehyde
Formula: C₈H₇BrO
Melting Pt: 33…37 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD04039890
CAS Number: 824-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR