4-Chloro-2-methylbenzyl alcohol 97%

Warning

H27554-06 H27554-03
AAH27554-03EA 70.2 USD
AAH27554-03 AAH27554-06
4-Chloro-2-methylbenzyl alcohol 97%
4-Chloro-2-methylbenzyl alcohol
Formula: C₈H₉ClO
MW: 156.61 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD06201140
CAS Number: 129716-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR