1-Hexyl-3-methylimidazolium tetrafluoroborate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: [HMIM] [BF4]1-n-Hexyl-3-methylimidazolium tetrafluoroborate

H27548-06 H27548-18
AAH27548-18EA 426.99 USD
AAH27548-18 AAH27548-06
1-Hexyl-3-methylimidazolium tetrafluoroborate 99%
1-Hexyl-3-methylimidazolium tetrafluoroborate
Formula: C₁₀H₁₉BF₄N₂
MW: 254.08 g/mol
Melting Pt: –81 °C
Density: 1.149
Storage Temperature: Ambient
MDL Number: MFCD03095459
CAS Number: 244193-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR