3-Chloro-2-methoxypyridine 95%

Warning

H27526-MD H27526-03
AAH27526-03EA 181.78 USD
AAH27526-03 AAH27526-MD
3-Chloro-2-methoxypyridine 95%
3-Chloro-2-methoxypyridine
Formula: C₆H₆ClNO
Storage Temperature: Ambient
MDL Number: MFCD03095227
CAS Number: 13472-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR