3-Chlor-2-methoxy-4-pyridineboronic acid pinacol ester 95%

Warning

H27513-MD H27513-03
AAH27513-03EA 250.86 USD
AAH27513-03 AAH27513-MD
3-Chlor-2-methoxy-4-pyridineboronic acid pinacol ester 95%
3-Chlor-2-methoxy-4-pyridineboronic acid pinacol ester
Formula: C₁₂H₁₇BClNO₃
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD06798258
CAS Number: 1073353-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR