2-Amino-3,5-diiodopyridine 95%

Warning

Synonyms: 3,5-Diiodopyridin-2-amine3,5-Diiodo-2-pyridylamine

H27511-03
AAH27511-03EA 114.33 USD
AAH27511-03
2-Amino-3,5-diiodopyridine 95%
2-Amino-3,5-diiodopyridine
Formula: C₅H₄I₂N₂
MW: 345.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02663852
CAS Number: 23597-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR