2,3-Dichloro-4-pyridineboronic acid pinacol ester 95%

Warning

Synonyms: 2,3-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

H27498-MD H27498-03
AAH27498-03EA 183.8 USD
AAH27498-03 AAH27498-MD
2,3-Dichloro-4-pyridineboronic acid pinacol ester 95%
2,3-Dichloro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₄BCl₂NO₂
MW: 273.95 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD06798257
CAS Number: 1073353-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR