2,4-Dibromopyrimidine 95%

Warning

H27433-06 H27433-03
AAH27433-03EA 316.64 USD
AAH27433-03 AAH27433-06
2,4-Dibromopyrimidine 95%
2,4-Dibromopyrimidine
Formula: C₄H₂Br₂N₂
MW: 237.88 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798232
CAS Number: 3921-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR