6-Chloro-2-pyridineboronic acid pinacol ester tech. 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
, 6-Chloropyridine-2-boronic acid pinacol ester
H27426-03
AAH27426-03EA
567.99
USD
AAH27426-03
6-Chloro-2-pyridineboronic acid pinacol ester tech. 90%, Technical Grade
6-Chloro-2-pyridineboronic acid pinacol ester
Formula:
C₁₁H₁₅BClNO₂ MW: 239.51 g/mol Melting Pt: 107…110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06798259 CAS Number: 652148-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...