5'-Chloro-2'-hydroxy-3'-nitroacetophenone 99%

Warning

Synonyms: 1-(5-Chloro-2-hydroxy-3-nitrophenyl)ethanone1-Chlor-3-ethanoyl-4-hydroxy-5-nitrobenzene

H27417-06 H27417-14
AAH27417-14EA 137.37 USD
AAH27417-14 AAH27417-06
5'-Chloro-2'-hydroxy-3'-nitroacetophenone 99%
5'-Chloro-2'-hydroxy-3'-nitroacetophenone
Formula: C₈H₆ClNO₄
MW: 215.59 g/mol
Melting Pt: 132…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00191921
CAS Number: 84942-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR