2,6-Dimethoxy-3-pyridineboronic acid pinacol ester 95%

Warning

H27282-MD H27282-03
AAH27282-MDEA 82.71 USD
AAH27282-MD AAH27282-03
2,6-Dimethoxy-3-pyridineboronic acid pinacol ester 95%
2,6-Dimethoxy-3-pyridineboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₄
Melting Pt: 39…42 °C
Storage Temperature: Ambient
MDL Number: MFCD03788240
CAS Number: 214360-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR